2-(Trifluoromethyl)quinoline 98%

Lieferant: Apollo Scientific
Achtung

PC0944-1G PC0944-5G
APOSPC0944-1GEA 274 EUR
APOSPC0944-1G APOSPC0944-5G
2-(Trifluoromethyl)quinoline 98%
2-(Trifluoromethyl)quinoline
Formel: C₁₀H₆F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD01862002
CAS-Nummer: 347-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR