1,1,2,3,3,4,4-Heptafluoro-1-butene 97%

Lieferant: Apollo Scientific
Achtung

PC0952-25G PC0952-5G
APOSPC0952-25GEA 425 EUR
APOSPC0952-25G APOSPC0952-5G
1,1,2,3,3,4,4-Heptafluoro-1-butene 97%
1,1,2,3,3,4,4-Heptafluoro-1-butene
Formel: C₄HF₇
Molecular Weight: 182,04 g/mol
Siedepunkt: 21 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00155824
CAS-Nummer: 680-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR