1,4-Difluoro-2-(methylsulphonyl)benzene 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,5-Difluorophenyl methyl sulphone, 1,4-Difluoro-2-(methylsulphonyl)benzene

PC0961-1G PC0961-5G
APOSPC0961-1GEA 219 EUR
APOSPC0961-1G APOSPC0961-5G
1,4-Difluoro-2-(methylsulphonyl)benzene 95%
1,4-Difluoro-2-(methylsulphonyl)benzene
Formel: C₇H₆F₂O₂S
Molecular Weight: 192.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06800566
CAS-Nummer: 236739-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR