Fmoc-D-4-fluorophe

Lieferant: Apollo Scientific

Synonyme: Fmoc-D-Phe(4-F)-OH, Fmoc-p-fluoro-D-Phe-OH, N-Fmoc-4-fluoro-D-phenylalanine

PC0967-1G PC0967-25G PC0967-5G
APOSPC0967-1GEA 26.1 EUR
APOSPC0967-1G APOSPC0967-25G APOSPC0967-5G
Fmoc-D-4-fluorophe
Fmoc-D-4-fluorophe
Formel: C₂₄H₂₀FNO₄
Molecular Weight: 405,425 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00235900
CAS-Nummer: 177966-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR