5-Phenyl-3-(trifluormethyl)-1H-pyrazol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Phenyl-3-(trifluoromethyl)pyrazole
PC10030-1G
PC10030-250MG
APOSPC10030-1GEA
45.1
EUR
APOSPC10030-1G
APOSPC10030-250MG
5-Phenyl-3-(trifluormethyl)-1H-pyrazol
5-Phenyl-3-(trifluormethyl)-1H-pyrazol
Formel:
C₁₀H₇F₃N₂ Molecular Weight: 212.18 g/mol Schmelzpunkt: 121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156064 CAS-Nummer: 4027-54-7 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...