Pentafluoro(trichloromethyl)benzene

Lieferant: Apollo Scientific
Achtung

Synonyme: (Trichloromethyl)pentafluorobenzene

PC10072-1G PC10072-5G PC10072-25G
APOSPC10072-1GEA 21.9 EUR
APOSPC10072-1G APOSPC10072-5G APOSPC10072-25G
Pentafluoro(trichloromethyl)benzene
Pentafluoro(trichloromethyl)benzene
Formel: C₇Cl₃F₅
Molecular Weight: 285,43 g/mol
Siedepunkt: 114…115 °C/48mmHg
Dichte: 1,6 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD01013630
CAS-Nummer: 778-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR