3-Iodo-1,1,1-trifluorobutane 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC10088-25G
PC10088-5G
APOSPC10088-25GEA
591
EUR
APOSPC10088-25G
APOSPC10088-5G
3-Iodo-1,1,1-trifluorobutane 97%
3-Iodo-1,1,1-trifluorobutane
Formel:
C₄H₆F₃I Molecular Weight: 237.99 g/mol Siedepunkt: 103.5 °C Flammpunkt: 40 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00077612 CAS-Nummer: 540-87-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...