2,4-Bis(trifluormethyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

PC10107-25G PC10107-100G PC10107-5G PC10107-1G
APOSPC10107-25GEA 176 EUR
APOSPC10107-25G APOSPC10107-100G APOSPC10107-5G APOSPC10107-1G
2,4-Bis(trifluormethyl)phenylboronsäure 98%
2,4-Bis(trifluormethyl)phenylboronsäure
Formel: C₈H₅BF₆O₂
Molecular Weight: 257,93 g/mol
Schmelzpunkt: 110…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01631349
CAS-Nummer: 153254-09-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR