2-(2-Fluorophenyl)-1-hydrazinecarbothioamide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2-Fluorophenyl)thiosemicarbazide

PC10118-1G PC10118-5G
APOSPC10118-1GEA 241 EUR
APOSPC10118-1G APOSPC10118-5G
2-(2-Fluorophenyl)-1-hydrazinecarbothioamide 95%
2-(2-Fluorophenyl)-1-hydrazinecarbothioamide
Formel: C₇H₈FN₃S
Molecular Weight: 185,22 g/mol
Schmelzpunkt: 198…201 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03791139
CAS-Nummer: 446275-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR