1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
Lieferant: Apollo Scientific
PC10123-1G
PC10123-500MG
PC10123-5G
APOSPC10123-1GEA
217
EUR
APOSPC10123-1G
APOSPC10123-500MG
APOSPC10123-5G
1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
Formel:
C₁₇H₁₃F₄N₃ Schmelzpunkt: 109…112 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791145 CAS-Nummer: 446275-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...