Methyl-2-amino-5-(4-fluorophenyl)-1,3-thiazole-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC10128-1G
PC10128-5G
APOSPC10128-1GEA
147
EUR
APOSPC10128-1G
APOSPC10128-5G
Methyl-2-amino-5-(4-fluorophenyl)-1,3-thiazole-4-carboxylate
Methyl-2-amino-5-(4-fluorophenyl)-1,3-thiazole-4-carboxylate
Formel:
C₁₁H₉FN₂O₂S Molecular Weight: 252.27 g/mol Schmelzpunkt: 202 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200913 CAS-Nummer: 886361-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...