3-Methyl-1-phenyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

Lieferant: Apollo Scientific

PC10130-10G PC10130-1G PC10130-5G
APOSPC10130-10GEA 519 EUR
APOSPC10130-10G APOSPC10130-1G APOSPC10130-5G
3-Methyl-1-phenyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
3-Methyl-1-phenyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine
Formel: C₁₇H₁₄F₃N₃
Schmelzpunkt: 104…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00140640
CAS-Nummer: 321385-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR