Ethyl-2-(5-amino-2-chloro-4-fluorophenoxy)acetate

Lieferant: Apollo Scientific
Achtung

PC10136-1G PC10136-5G
APOSPC10136-1GEA 292 EUR
APOSPC10136-1G APOSPC10136-5G
Ethyl-2-(5-amino-2-chloro-4-fluorophenoxy)acetate
Ethyl-2-(5-amino-2-chloro-4-fluorophenoxy)acetate
Formel: C₁₀H₁₁ClFNO₃
Molecular Weight: 247.66 g/mol
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425716
CAS-Nummer: 91920-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR