4-(3-Fluorpropoxy)benzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
PC10138-10MG
PC10138-100MG
APOSPC10138-10MGEA
21.9
EUR
APOSPC10138-10MG
APOSPC10138-100MG
4-(3-Fluorpropoxy)benzaldehyd
4-(3-Fluoropropoxy)benzaldehyde
Formel:
C₁₀H₁₁FO₂ Siedepunkt: 92…94 °C/0.1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD02090999 CAS-Nummer: 400825-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...