Methyl-2-[(3-fluorobenzyl)sulphanyl]benzenecarboxylate
Lieferant: Apollo Scientific
PC10144-1G
PC10144-500MG
PC10144-5G
APOSPC10144-1GEA
217
EUR
APOSPC10144-1G
APOSPC10144-500MG
APOSPC10144-5G
Methyl-2-[(3-fluorobenzyl)sulphanyl]benzenecarboxylate
Methyl-2-[(3-fluorobenzyl)sulphanyl]benzenecarboxylate
Formel:
C₁₅H₁₃FO₂S Molecular Weight: 276,33 g/mol Schmelzpunkt: 70…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001233 CAS-Nummer: 385383-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...