4-[(2-Fluorobenzyl)oxy]benzenecarbonitrile

Lieferant: Apollo Scientific

Synonyme: 4-(2-Fluorobenzyloxy)benzonitrile

PC10149-10G PC10149-1G PC10149-5G
APOSPC10149-10GEA 372 EUR
APOSPC10149-10G APOSPC10149-1G APOSPC10149-5G
4-[(2-Fluorobenzyl)oxy]benzenecarbonitrile
4-[(2-Fluorobenzyl)oxy]benzenecarbonitrile
Formel: C₁₄H₁₀FNO
Molecular Weight: 227,24 g/mol
Schmelzpunkt: 80…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681655
CAS-Nummer: 852690-97-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR