2-Chloro-4-(trifluoromethyl)pyridine-3-carboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4-(trifluoromethyl)nicotinamide

PC10156-1G PC10156-500MG
APOSPC10156-1GEA 386 EUR
APOSPC10156-1G APOSPC10156-500MG
2-Chloro-4-(trifluoromethyl)pyridine-3-carboxamide
2-Chloro-4-(trifluoromethyl)pyridine-3-carboxamide
Formel: C₇H₄ClF₃N₂O
Molecular Weight: 224.57 g/mol
Schmelzpunkt: 187…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04039215
CAS-Nummer: 886762-28-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR