3-[4-(3-Fluoropropoxy)phenyl]acrylic acid

Lieferant: Apollo Scientific

PC10173-1G PC10173-250MG
APOSPC10173-1GEA 364 EUR
APOSPC10173-1G APOSPC10173-250MG
3-[4-(3-Fluoropropoxy)phenyl]acrylic acid
3-[4-(3-Fluoropropoxy)phenyl]acrylic acid
Formel: C₁₂H₁₃FO₃
Schmelzpunkt: 176…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02082811
CAS-Nummer: 695186-85-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR