[4-(3-Fluoropropoxy)phenyl]methanol

Lieferant: Apollo Scientific
Achtung

PC10174-1G PC10174-500MG PC10174-5G
APOSPC10174-1GEA 533 EUR
APOSPC10174-1G APOSPC10174-500MG APOSPC10174-5G
[4-(3-Fluoropropoxy)phenyl]methanol
[4-(3-Fluoropropoxy)phenyl]methanol
Formel: C₁₀H₁₃FO₂
Molecular Weight: 184.21 g/mol
Schmelzpunkt: 52…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091005
CAS-Nummer: 449778-58-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR