6-Methyl-4-trifluoromethyl-2(1H)-pyridone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one
PC10177-1G
PC10177-5G
APOSPC10177-1GEA
183
EUR
APOSPC10177-1G
APOSPC10177-5G
6-Methyl-4-trifluoromethyl-2(1H)-pyridone
6-Methyl-4-trifluoromethyl-2(1H)-pyridone
Formel:
C₇H₆F₃NO Molecular Weight: 177,126 g/mol Schmelzpunkt: 124…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042479 CAS-Nummer: 22123-19-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...