2-Amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile

Lieferant: Apollo Scientific

PC10180-1G PC10180-500MG
APOSPC10180-1GEA 799 EUR
APOSPC10180-1G APOSPC10180-500MG
2-Amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile
2-Amino-6-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile
Formel: C₁₄H₁₀ClF₃N₄S
Molecular Weight: 358.77 g/mol
Schmelzpunkt: 199…202 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00180796
CAS-Nummer: 321430-41-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR