1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbohydrazide
Lieferant: Apollo Scientific
PC10181-1G
PC10181-500MG
PC10181-5G
APOSPC10181-1GEA
464
EUR
APOSPC10181-1G
APOSPC10181-500MG
APOSPC10181-5G
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbohydrazide
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbohydrazide
Formel:
C₁₂H₁₄ClF₃N₄O Molecular Weight: 322.72 g/mol Schmelzpunkt: 198…201 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00172276 CAS-Nummer: 321430-54-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...