4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methyl}benzenol

Lieferant: Apollo Scientific

PC10182-1G PC10182-500MG
APOSPC10182-1GEA 533 EUR
APOSPC10182-1G APOSPC10182-500MG
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methyl}benzenol
4-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]methyl}benzenol
Formel: C₁₃H₉ClF₃NO
Molecular Weight: 287,67 g/mol
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01316641
CAS-Nummer: 321432-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR