3-[2-(4-Fluorophenoxy)phenyl]acrylic acid

Lieferant: Apollo Scientific

PC10185-1G PC10185-5G
APOSPC10185-1GEA 434 EUR
APOSPC10185-1G APOSPC10185-5G
3-[2-(4-Fluorophenoxy)phenyl]acrylic acid
3-[2-(4-Fluorophenoxy)phenyl]acrylic acid
Formel: C₁₅H₁₁FO₃
Schmelzpunkt: 205…207 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091006
CAS-Nummer: 338393-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR