3-[2-(3,4-Difluorophenoxy)phenyl]acrylic acid

Lieferant: Apollo Scientific

PC10186-1G PC10186-500MG
APOSPC10186-1GEA 671 EUR
APOSPC10186-1G APOSPC10186-500MG
3-[2-(3,4-Difluorophenoxy)phenyl]acrylic acid
3-[2-(3,4-Difluorophenoxy)phenyl]acrylic acid
Formel: C₁₅H₁₀F₂O₃
Molecular Weight: 276.24 g/mol
Schmelzpunkt: 198…200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091009
CAS-Nummer: 338393-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR