3-[2-(4-Chloro-3-fluorophenoxy)phenyl]acrylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: trans-2-(4-Chloro-3-fluorophenoxy)cinnamic acid

PC10187-1G PC10187-500MG
APOSPC10187-1GEA 372 EUR
APOSPC10187-1G APOSPC10187-500MG
3-[2-(4-Chloro-3-fluorophenoxy)phenyl]acrylic acid
3-[2-(4-Chloro-3-fluorophenoxy)phenyl]acrylic acid
Formel: C₁₅H₁₀ClFO₃
Molecular Weight: 292.7 g/mol
Schmelzpunkt: 197…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02091010
CAS-Nummer: 449778-67-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR