2,2,2-Trifluoro-N-(6-oxo-5,6-dihydro-4H-cyclopenta[b]thiophen-4-yl)acetamide

Lieferant: Apollo Scientific

PC10196-1G PC10196-500MG PC10196-5G
APOSPC10196-1GEA 405 EUR
APOSPC10196-1G APOSPC10196-500MG APOSPC10196-5G
2,2,2-Trifluoro-N-(6-oxo-5,6-dihydro-4H-cyclopenta[b]thiophen-4-yl)acetamide
2,2,2-Trifluoro-N-(6-oxo-5,6-dihydro-4H-cyclopenta[b]thiophen-4-yl)acetamide
Formel: C₉H₆F₃NO₂S
Molecular Weight: 249,21 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03012801
CAS-Nummer: 108046-14-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR