2-Amino-5-(4-fluorphenyl)-4-thiazolcarbohydrazid
Lieferant: Apollo Scientific
PC10203-1G
PC10203-500MG
PC10203-5G
APOSPC10203-1GEA
217
EUR
APOSPC10203-1G
APOSPC10203-500MG
APOSPC10203-5G
2-Amino-5-(4-fluorphenyl)-4-thiazolcarbohydrazid
2-Amino-5-(4-fluorphenyl)-4-thiazolcarbohydrazid
Formel:
C₁₀H₉FN₄OS Schmelzpunkt: 174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200939 CAS-Nummer: 886361-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...