3-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
PC10217-1G
PC10217-250MG
APOSPC10217-1GEA
290
EUR
APOSPC10217-1G
APOSPC10217-250MG
3-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1H-pyrazole
3-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}-1H-pyrazole
Formel:
C₁₆H₁₂ClFN₂O Molecular Weight: 302.74 g/mol Schmelzpunkt: 125…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425750 CAS-Nummer: 451485-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...