Methyl-1-[(4-fluorophenyl)sulphonyl]-4-piperidinecarboxylate

Lieferant: Apollo Scientific

PC10218-1G PC10218-500MG PC10218-5G
APOSPC10218-1GEA 217 EUR
APOSPC10218-1G APOSPC10218-500MG APOSPC10218-5G
Methyl-1-[(4-fluorophenyl)sulphonyl]-4-piperidinecarboxylate
Methyl-1-[(4-fluorophenyl)sulphonyl]-4-piperidinecarboxylate
Formel: C₁₃H₁₆FNO₄S
Molecular Weight: 301.34 g/mol
Schmelzpunkt: 90…91 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02736084
CAS-Nummer: 349624-64-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR