3-(4-Fluorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid

Lieferant: Apollo Scientific

PC10224-1G PC10224-500MG PC10224-5G
APOSPC10224-1GEA 172 EUR
APOSPC10224-1G APOSPC10224-500MG APOSPC10224-5G
3-(4-Fluorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
3-(4-Fluorophenyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Formel: C₁₁H₉F₄NO₃
Molecular Weight: 279.19 g/mol
Schmelzpunkt: 135…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01366669
CAS-Nummer: 117291-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR