Methyl-3-(2-amino-4-fluorophenoxy)-2-thiophenecarboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC10227-1G
PC10227-250MG
APOSPC10227-1GEA
115
EUR
APOSPC10227-1G
APOSPC10227-250MG
Methyl-3-(2-amino-4-fluorophenoxy)-2-thiophenecarboxylate
Methyl-3-(2-amino-4-fluorophenoxy)-2-thiophenecarboxylate
Formel:
C₁₂H₁₀FNO₃S Molecular Weight: 267,28 g/mol Schmelzpunkt: 106…108 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04117805 CAS-Nummer: 886360-59-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...