Methyl-4-hydroxy-1-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyrrolidinecarboxylate
Lieferant: Apollo Scientific
PC10232-1G
PC10232-500MG
APOSPC10232-1GEA
666
EUR
APOSPC10232-1G
APOSPC10232-500MG
Methyl-4-hydroxy-1-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyrrolidinecarboxylate
Methyl-4-hydroxy-1-{[3-(trifluoromethyl)anilino]carbonyl}-2-pyrrolidinecarboxylate
Formel:
C₁₄H₁₅F₃N₂O₄ Molecular Weight: 332,28 g/mol Schmelzpunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202782 CAS-Nummer: 318247-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...