1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Amino-2,2,2-trifluoroacetophenone, 1-(4-Aminophenyl)-2,2,2-trifluoroethanone

PC10234-1G PC10234-5G
APOSPC10234-1GEA 130 EUR
APOSPC10234-1G APOSPC10234-5G
1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone
1-(4-Aminophenyl)-2,2,2-trifluoro-1-ethanone
Formel: C₈H₆F₃NO
Molecular Weight: 189,14 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Kühlschrank
MDL: MFCD05663790
CAS-Nummer: 23516-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR