Ethyl-2-oxo-4-(trifluoromethyl)-2,3-dihydro-1,3-thiazole-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
PC10242-1G
PC10242-250MG
APOSPC10242-1GEA
719
EUR
APOSPC10242-1G
APOSPC10242-250MG
Ethyl-2-oxo-4-(trifluoromethyl)-2,3-dihydro-1,3-thiazole-5-carboxylate
Ethyl-2-oxo-4-(trifluoromethyl)-2,3-dihydro-1,3-thiazole-5-carboxylate
Formel:
C₇H₆F₃NO₃S Molecular Weight: 241,19 g/mol Schmelzpunkt: 122…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425775 CAS-Nummer: 72850-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...