5-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Trifluoromethyl)benzo[b]thiophene-2-carboxylic acid
PC10245-1G
PC10245-10G
PC10245-5G
APOSPC10245-1GEA
40.6
EUR
APOSPC10245-1G
APOSPC10245-10G
APOSPC10245-5G
5-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
5-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid
Formel:
C₁₀H₅F₃O₂S Molecular Weight: 246,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03012933 CAS-Nummer: 244126-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...