2'-(4-Fluorbenzyloxy)acetophenon
Lieferant: Apollo Scientific
PC10254-1G
PC10254-5G
APOSPC10254-1GEA
137
EUR
APOSPC10254-1G
APOSPC10254-5G
2'-(4-Fluorbenzyloxy)acetophenon
2'-(4-Fluorbenzyloxy)acetophenon
Formel:
C₁₅H₁₃FO₂ Schmelzpunkt: 56…58 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001269 CAS-Nummer: 400878-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...