3-[(4-Fluorobenzyl)oxy]-2-methyl-4(1H)-pyridinone

Lieferant: Apollo Scientific

PC10260-1G PC10260-500MG PC10260-5G
APOSPC10260-1GEA 217 EUR
APOSPC10260-1G APOSPC10260-500MG APOSPC10260-5G
3-[(4-Fluorobenzyl)oxy]-2-methyl-4(1H)-pyridinone
3-[(4-Fluorobenzyl)oxy]-2-methyl-4(1H)-pyridinone
Formel: C₁₃H₁₂FNO₂
Molecular Weight: 233,24 g/mol
Schmelzpunkt: 129…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00215663
CAS-Nummer: 338965-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR