2-Phenethyl-1-(trifluoromethyl)-1,2,5,6-tetrahydro-4H-pyrrolo[3,2,1-ij]quinolin-1-ol
Lieferant: Apollo Scientific
PC10265-1G
PC10265-500MG
APOSPC10265-1GEA
1060
EUR
APOSPC10265-1G
APOSPC10265-500MG
2-Phenethyl-1-(trifluoromethyl)-1,2,5,6-tetrahydro-4H-pyrrolo[3,2,1-ij]quinolin-1-ol
2-Phenethyl-1-(trifluoromethyl)-1,2,5,6-tetrahydro-4H-pyrrolo[3,2,1-ij]quinolin-1-ol
Formel:
C₂₀H₂₀F₃NO Schmelzpunkt: 76…78 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425797 CAS-Nummer: 439095-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...