1-Fluoro-4-[(4-nitrophenyl)sulphanyl]benzene
Lieferant: Apollo Scientific
Synonyme:
(4-Fluorophenyl)(4-nitrophenyl)sulphane, 4-[(4-Fluorophenyl)thio]nitrobenzene
PC10269-1G
PC10269-500MG
PC10269-5G
APOSPC10269-1GEA
147
EUR
APOSPC10269-1G
APOSPC10269-500MG
APOSPC10269-5G
1-Fluoro-4-[(4-nitrophenyl)sulphanyl]benzene
1-Fluoro-4-[(4-nitrophenyl)sulphanyl]benzene
Formel:
C₁₂H₈FNO₂S Molecular Weight: 249,27 g/mol Schmelzpunkt: 92…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04117815 CAS-Nummer: 2438-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...