6-Methyl-2-(2,2,2-trifluoro-1-hydroxyethyl)-3-pyridinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Hydroxy-2-(1-hydroxy-2,2,2-trifluoroethyl)-6-methylpyridine
PC10280-1G
PC10280-5G
APOSPC10280-1GEA
168
EUR
APOSPC10280-1G
APOSPC10280-5G
6-Methyl-2-(2,2,2-trifluoro-1-hydroxyethyl)-3-pyridinol
6-Methyl-2-(2,2,2-trifluoro-1-hydroxyethyl)-3-pyridinol
Formel:
C₈H₈F₃NO₂ Molecular Weight: 207,15 g/mol Schmelzpunkt: 138…141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03425809 CAS-Nummer: 344776-71-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...