1,1,1-Trifluoro-3-{4-[3-(trifluoromethyl)phenyl]piperazino}-2-propanol

Lieferant: Apollo Scientific

PC10291-1G PC10291-500MG
APOSPC10291-1GEA 924 EUR
APOSPC10291-1G APOSPC10291-500MG
1,1,1-Trifluoro-3-{4-[3-(trifluoromethyl)phenyl]piperazino}-2-propanol
1,1,1-Trifluoro-3-{4-[3-(trifluoromethyl)phenyl]piperazino}-2-propanol
Formel: C₁₄H₁₆F₆N₂O
Molecular Weight: 342.29 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01871732
CAS-Nummer: 453557-82-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR