Pentafluorphenyl-2-(trifluormethyl)benzolsulfonat

Lieferant: Apollo Scientific

PC10300-1G PC10300-500MG PC10300-5G
APOSPC10300-1GEA 275 EUR
APOSPC10300-1G APOSPC10300-500MG APOSPC10300-5G
Pentafluorphenyl-2-(trifluormethyl)benzolsulfonat
Pentafluorphenyl-2-(trifluormethyl)benzolsulfonat
Formel: C₁₃H₄F₈O₃S
Schmelzpunkt: 79…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05975148
CAS-Nummer: 886361-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR