3-[(2,3,4,5,6-Pentafluorophenoxy)sulphonyl]benzenecarboxylic acid

Lieferant: Apollo Scientific

Synonyme: 3-[(2,3,4,5,6-Pentafluorophenoxy)sulphonyl]benzoic acid

PC10301-1G PC10301-500MG PC10301-5G
APOSPC10301-1GEA 275 EUR
APOSPC10301-1G APOSPC10301-500MG APOSPC10301-5G
3-[(2,3,4,5,6-Pentafluorophenoxy)sulphonyl]benzenecarboxylic acid
3-[(2,3,4,5,6-Pentafluorophenoxy)sulphonyl]benzenecarboxylic acid
Formel: C₁₃H₅F₅O₅S
Molecular Weight: 368.24 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05975151
CAS-Nummer: 886361-31-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR