Pentafluorophenyl-2-thiophensulfonat
Lieferant: Apollo Scientific
PC10314-1G
PC10314-5G
APOSPC10314-1GEA
139
EUR
APOSPC10314-1G
APOSPC10314-5G
Pentafluorophenyl-2-thiophensulfonat
Pentafluorophenyl-2-thiophensulfonat
Formel:
C₁₀H₃F₅O₃S₂ Schmelzpunkt: 90…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD05975124 CAS-Nummer: 663175-91-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...