Pentafluorphenyl-3-methylbenzolsulfonat

Lieferant: Apollo Scientific

PC10320-1G PC10320-500MG PC10320-5G
APOSPC10320-1GEA 275 EUR
APOSPC10320-1G APOSPC10320-500MG APOSPC10320-5G
Pentafluorphenyl-3-methylbenzolsulfonat
Pentafluorphenyl-3-methylbenzolsulfonat
Formel: C₁₃H₇F₅O₃S
Schmelzpunkt: 49…51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05975125
CAS-Nummer: 885950-34-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR