4-Cyan-3-fluorphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyano-3-fluorobenzeneboronic acid

PC10333-1G PC10333-25G PC10333-5G PC10333-10G
APOSPC10333-1GEA 21.9 EUR
APOSPC10333-1G APOSPC10333-25G APOSPC10333-5G APOSPC10333-10G
4-Cyan-3-fluorphenylboronsäure
4-Cyan-3-fluorphenylboronsäure
Formel: C₇H₅BFNO₂
Molecular Weight: 164.93 g/mol
Schmelzpunkt: 362…366 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411549
CAS-Nummer: 843663-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR