3-(2-Thienyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Lieferant: Apollo Scientific
PC10348-1G
PC10348-500MG
PC10348-5G
APOSPC10348-1GEA
217
EUR
APOSPC10348-1G
APOSPC10348-500MG
APOSPC10348-5G
3-(2-Thienyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
3-(2-Thienyl)-3-[(2,2,2-trifluoroacetyl)amino]propanoic acid
Formel:
C₉H₈F₃NO₃S Schmelzpunkt: 154…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001310 CAS-Nummer: 115957-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...