5-Fluoroisophthalonitrile 99+%

Lieferant: Apollo Scientific
Achtung

PC10349-5G PC10349-10G
APOSPC10349-5GEA 27.7 EUR
APOSPC10349-5G APOSPC10349-10G
5-Fluoroisophthalonitrile 99+%
5-Fluoroisophthalonitrile
Formel: C₈H₃FN₂
Molecular Weight: 146.12 g/mol
Schmelzpunkt: 109…111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05663831
CAS-Nummer: 453565-55-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR