3-Fluor-2-methylphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Fluoro-2-methylphenyl)boronic acid, 3-Fluoro-2-methylbenzeneboronic acid
PC10380-1G
PC10380-5G
APOSPC10380-1GEA
50.9
EUR
APOSPC10380-1G
APOSPC10380-5G
3-Fluor-2-methylphenylboronsäure 98%
3-Fluor-2-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153.95 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368679 CAS-Nummer: 163517-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...